FRO048767975 Frontier Scientific Services Inc.

4-[(5-Chloro-1,3-Dimethyl-1H-Pyrazol-4-Yl)Sulfonyl]Morpholine

  • MDL number MFCD00113902
  • Molecular Weight : 279.7472

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart