FRO048768018 Frontier Scientific Services Inc.

(1H-Imidazol-2-Yl-Methyl)(Methyl)Amine Dihydrochloride

  • MDL number MFCD07781074
  • Molecular Weight : 184.067103

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Dihydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart