FRO048768023 Frontier Scientific Services Inc.

Methyl-(1-Pyridin-4-Yl-Ethyl)-Amine Oxalate

  • MDL number MFCD09048938
  • Molecular Weight : 181.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Oxalate
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart