FRO048768148 Frontier Scientific Services Inc.

2-Amino-3-(2,5-DiMethyl-Benzyl-Oxy)-6-Methylpyridine

  • MDL number MFCD01313008
  • Cas Number : 300665-41-2
  • Molecular Weight : 242.3235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart