FRO048768162 Frontier Scientific Services Inc.

2-Chloroethylamine Hydrochloride

  • MDL number MFCD00012887
  • Cas Number : 870-24-6
  • Molecular Weight : 115.9898

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart