FRO048768273 Frontier Scientific Services Inc.

4'-Methoxy-Biphenyl-2-Yl-Amine

  • MDL number MFCD03424652
  • Molecular Weight : 199.2547

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart