FRO048768394 Frontier Scientific Services Inc.

2,6 Dibenzylphenol

  • MDL number MFCD02683215
  • Cas Number : 47157-01-7
  • Molecular Weight : 274.3659

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart