FRO048768427 Frontier Scientific Services Inc.

4-Hex-1-Ynyl-Benzaldehyde

  • MDL number MFCD00276593
  • Cas Number : 175203-90-4
  • Molecular Weight : 186.2559

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart