FRO048768438 Frontier Scientific Services Inc.

4-N-HeptYl-Oxybenzoyl Chloride

  • MDL number MFCD00000691
  • Cas Number : 40782-54-5
  • Molecular Weight : 254.7593

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart