FRO048768522 Frontier Scientific Services Inc.

4-[2-(Piperidin-4-Yl)Ethyl]Piperidine Dihydrochloride

  • MDL number MFCD00012774
  • Cas Number : 80997-86-0
  • Molecular Weight : 269.254397

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Dihydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart