FRO048768646 Frontier Scientific Services Inc.

N1-[3,5-di(trifluoromethyl)phenyl]-2-[(2-aminophenyl)thio]-2-phenylacetamid e

  • MDL number MFCD00124787
  • Molecular Weight : 470.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart