FRO048769037 Frontier Scientific Services Inc.

2-(2-Amino-2-Methyl-Propylamino)-2-Methyl-Propan-1-Ol

  • MDL number MFCD00025544
  • Molecular Weight : 160.2614

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart