FRO048769077 Frontier Scientific Services Inc.

2-Ethylsulfanyl-6-Methyl-Pyrimidine-4-Thiol

  • MDL number MFCD00023248
  • Cas Number : 6967-58-4
  • Molecular Weight : 186.2992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart