FRO048769099 Frontier Scientific Services Inc.

3-METHYL-1-METHYLSULFANYL-BUTANE

  • MDL number MFCD00026521
  • Molecular Weight : 118.2425

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart