FRO048769146 Frontier Scientific Services Inc.

3-Morpholinopropiophenone Hydrochloride

  • MDL number MFCD00035332
  • Cas Number : 1020-16-2
  • Molecular Weight : 255.7459

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart