FRO048769159 Frontier Scientific Services Inc.

3-Benzylidene-Dihydro-Furan-2-One

  • MDL number MFCD00022487
  • Molecular Weight : 174.2012

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart