FRO048769196 Frontier Scientific Services Inc.

2-Hydroxy-Propane-2-Sulfonic Acid Sodium Salt

  • MDL number MFCD00031395
  • Molecular Weight : 162.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Sodium Salt
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart