FRO048956887 Frontier Scientific Services Inc.

L-Glutamic acid 1-(4-nitroanilide)

  • MDL number MFCD00038111
  • Cas Number : 24032-35-7
  • Molecular Weight : 266.25578

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart