FRO048956899 Frontier Scientific Services Inc.

N'-hydroxy-2-(propylsulfonyl)ethanimidamide

  • MDL number MFCD02678227
  • Molecular Weight : 180.22699

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart