FRO068581683 Frontier Scientific Services Inc.

7-[(Chlorocarbonyl)Methoxy]-4-Methylcoumarin

  • MDL number MFCD00143299
  • Cas Number : 91454-65-8
  • Molecular Weight : 252.6561

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart