FRO068581694 Frontier Scientific Services Inc.

[2-Fluoro-6-(4-Fluoro-Phenoxy)-Benzyl]-Methyl-Amine

  • MDL number MFCD07368318
  • Molecular Weight : 249.2626

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart