FRO068581709 Frontier Scientific Services Inc.

4-Ethyl-4-Formylhexanenitrile

  • MDL number MFCD00047654
  • Cas Number : 2938-69-4
  • Molecular Weight : 153.221497

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart