FRO068581726 Frontier Scientific Services Inc.

2-(2-Formyl-1H-Pyrrol-1-Yl)-4-Methoxynicotinonitrile

  • MDL number MFCD00975451
  • Molecular Weight : 227.2244

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart