FRO068581755 Frontier Scientific Services Inc.

3,5-Dichloro-4-Methoxybenzoyl Chloride

  • MDL number MFCD02063082
  • Cas Number : 29568-76-1
  • Molecular Weight : 239.483002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart