FRO068581776 Frontier Scientific Services Inc.

(2,4-DICHLORO-6-METHYL-PHENOXY)-ACETYL CHLORIDE

  • MDL number MFCD02258047
  • Molecular Weight : 253.5139

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart