FRO068581850 Frontier Scientific Services Inc.

2-(1H-Pyrazol-3-Yl)-1,3-Benzothiazole Hydrochloride

  • MDL number MFCD01934517
  • Cas Number : 256414-72-9
  • Molecular Weight : 237.701

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart