FRO068581979 Frontier Scientific Services Inc.

5-([(4-Chlorobenzoyl)Amino]Methyl)Thiophene-2-Sulfonyl Chloride

  • MDL number MFCD00084934
  • Molecular Weight : 350.2444

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart