FRO068582019 Frontier Scientific Services Inc.

(2S)-2-Amino-N-Methylpropanamide Hydrochloride

  • MDL number MFCD00237739
  • Cas Number : 61275-22-7
  • Molecular Weight : 138.596002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart