FRO068848746 Frontier Scientific Services Inc.

3-(2-Amino-Pyridin-3-Yl)-Acrylic Acid

  • MDL number MFCD06205691
  • Molecular Weight : 164.1652

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart