FRO068848760 Frontier Scientific Services Inc.

3-[4-(2,2-Dimethyl-Propionylamino)-Pyridin-3-Yl]-Acrylic Acid Ethyl Ester

  • MDL number MFCD07781148
  • Molecular Weight : 276.3383

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart