FRO068848761 Frontier Scientific Services Inc.

Ethyl (2E)-3-(4-Aminopyridin-3-Yl)Prop-2-Enoate

  • MDL number MFCD01941262
  • Cas Number : 104830-07-1
  • Molecular Weight : 192.214493

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart