FRO068848814 Frontier Scientific Services Inc.

3-(5-Bromo-2-Methoxy-Pyridin-3-Yl)-Acrylic Acid Ethyl Ester

  • MDL number MFCD07781177
  • Molecular Weight : 286.1272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart