FRO068848827 Frontier Scientific Services Inc.

5'-Iodo-3,4,5,6-Tetrahydro-2H-[1,2']Bipyridinyl

  • MDL number MFCD07781187
  • Molecular Weight : 288.1329

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart