FRO068848830 Frontier Scientific Services Inc.

3-(6-Pyrrolidin-1-Yl-Pyridin-3-Yl)-Acrylic Acid Ethyl Ester

  • MDL number MFCD07781189
  • Molecular Weight : 246.3118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart