FRO068848831 Frontier Scientific Services Inc.

3-(3,4,5,6-Tetrahydro-2H-[1,2']Bipyridinyl-5'-Yl)-Acrylic Acid Ethyl Ester

  • MDL number MFCD07781190
  • Molecular Weight : 260.3389

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart