FRO087887140 Frontier Scientific Services Inc.

4-Hydrazino-5-Methylthieno[2,3-D]Pyrimidine

  • MDL number MFCD00205195
  • Cas Number : 19786-56-2
  • Molecular Weight : 180.2326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart