FRO087887155 Frontier Scientific Services Inc.

4-(4-Aminobenzyl)-Thiomorpholine-1,1-Dioxide

  • MDL number MFCD03407300
  • Cas Number : 263339-24-8
  • Molecular Weight : 240.322006

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart