FRO087887157 Frontier Scientific Services Inc.

4-(4-Methoxyphenyl)-1,2,3-Thiadiazol-5-Amine

  • MDL number MFCD03407307
  • Cas Number : 115842-95-0
  • Molecular Weight : 207.2556

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart