FRO087887395 Frontier Scientific Services Inc.

1-THIOPHEN-2-YL-PROPAN-2-OL

  • MDL number MFCD00022533
  • Molecular Weight : 142.2212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart