FRO087887439 Frontier Scientific Services Inc.

3,4-Dimethylphenylhydrazine Hydrochloride

  • MDL number MFCD00052270
  • Molecular Weight : 172.6582

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart