FRO087887517 Frontier Scientific Services Inc.

(6-Morpholino-3-Pyridinyl)Methanol

  • MDL number MFCD03086115
  • Molecular Weight : 194.2353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart