FRO091856342 Frontier Scientific Services Inc.

4-[4-(4-Methoxyphenyl)-1H-Pyrazol-1-Yl]Piperidine

  • MDL number MFCD08060996
  • Molecular Weight : 257.330902

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart