FRO091856343 Frontier Scientific Services Inc.

4-[4-(4-Chloro-Phenyl)-Pyrazol-1-Yl]-Piperidine

  • MDL number MFCD08060997
  • Molecular Weight : 261.7567

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart