FRO091856351 Frontier Scientific Services Inc.

4-(2-Chloro-Phenyl)-Tetrahydro-Pyran-4-Carbaldehyde

  • MDL number MFCD08061003
  • Molecular Weight : 224.6892

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart