FRO091856364 Frontier Scientific Services Inc.

5-Fluoro-2-(4-Fluorophenoxy)Benzaldehyde

  • MDL number MFCD08060960
  • Molecular Weight : 234.198196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart