FRO091856389 Frontier Scientific Services Inc.

1-(8-Aza-Bicyclo[3.2.1]Oct-3-Yl)-1H-Benzoimidazole

  • MDL number MFCD08064259
  • Molecular Weight : 227.3117

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart