FRO091856405 Frontier Scientific Services Inc.

[4-(2-Fluoro-Phenoxymethyl)-Phenyl]-Acetic Acid

  • MDL number MFCD08060966
  • Molecular Weight : 260.2675

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart