FRO091856433 Frontier Scientific Services Inc.

2-Hydroxy-3,4-Dimethoxy-6-Methyl-Benzaldehyde

  • MDL number MFCD00075802
  • Cas Number : 22383-86-4
  • Molecular Weight : 196.199905

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart