FRO091856467 Frontier Scientific Services Inc.

1-Acetyl-N-Isobutylpyrrolidin-3-Amine

  • MDL number MFCD08061023
  • Molecular Weight : 184.2837

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart