FRO091856472 Frontier Scientific Services Inc.

N,N-Dimethyl-2-Piperazin-1-Yl-Nicotinamide

  • MDL number MFCD08061072
  • Molecular Weight : 234.3035

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart